APO414281672 Apollo Scientific Ltd

Ethyl 1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD18262320
  • Cas Number : 852228-09-2
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-1-methyl-5-trifluoromethyl-1h-pyrazole-3-carboxylate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/ethyl-1-methyl-5-trifluoromethyl-1h-pyrazole-3-carboxylate
Availability details for
Quantity
  • Availability
  • Add to Cart