APO414281714 Apollo Scientific Ltd

4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one

  • MDL number MFCD18384918
  • Cas Number : 1007578-99-5
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-bromophenyl-24-dihydro-3h-124-triazol-3-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-bromophenyl-24-dihydro-3h-124-triazol-3-one
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart