APO414281731 Apollo Scientific Ltd

2-Ethyl-1,3-benzoxazol-6-amine

  • MDL number MFCD03791184
  • Cas Number : 855423-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-ethyl-13-benzoxazol-6-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-ethyl-13-benzoxazol-6-amine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart