APO414281769 Apollo Scientific Ltd

1-Chloro-3-(morpholin-4-yl)propane

  • MDL number MFCD00039714
  • Cas Number : 7357-67-7
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-chloropropyl)morpholine
density 1.08
Inchi Code 1S/C7H14ClNO/c8-2-1-3-9-4-6-10-7-5-9/h1-7H2
Inchi Key PIAZYBLGBSMNLX-UHFFFAOYSA-N

Description

Synonym 1-Chloro-3-morpholinopropane
Density 1.08
Flash Point 115/25
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-chloro-3-morpholin-4-ylpropane

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-chloro-3-morpholin-4-ylpropane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart