APO414281779 Apollo Scientific Ltd

Ethyl 2,6-dichloropyridine-3-carboxylate

  • MDL number MFCD09757510
  • Cas Number : 58584-86-4
  • Molecular Weight : 220.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 50-51
Iupac Name ethyl 2,6-dichloronicotinate
Inchi Code 1S/C8H7Cl2NO2/c1-2-13-8(12)5-3-4-6(9)11-7(5)10/h3-4H,2H2,1H3
Inchi Key KKYBVLDQTWQETN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 50-51
Product Link https://store.apolloscientific.co.uk/product/ethyl-26-dichloropyridine-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-26-dichloropyridine-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart