APO414281818 Apollo Scientific Ltd

Dimesna

  • MDL number MFCD00868218
  • Cas Number : 16208-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >274 (dec.)
Synonym Disodium 2,2'-dithiobisethane sulfonate, Sodium 2,2'-disulfanediyldiethanesulfonate
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/dimesna

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/dimesna
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart