APO414281871 Apollo Scientific Ltd

4-Pyridin-2-ylpyrimidin-2-amine

  • MDL number MFCD00138711
  • Cas Number : 66521-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-pyridin-2-ylpyrimidin-2-amine

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-pyridin-2-ylpyrimidin-2-amine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart