APO414281882 Apollo Scientific Ltd

Ethyl 4-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD12498713
  • Cas Number : 1171919-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50638

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50638
pictograms GHS07
hstatements H302;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart