APO414281886 Apollo Scientific Ltd

5-Bromo-4-hydroxy-6-(trifluoromethyl)pyrimidine

  • Molecular Weight : 242.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 225-227
Iupac Name 5-bromo-6-(trifluoromethyl)-4-pyrimidinol
Inchi Code 1S/C5H2BrF3N2O/c6-2-3(5(7,8)9)10-1-11-4(2)12/h1H,(H,10,11,12)
Inchi Key GUMRBYBFTLXIKZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 225-227

Safety Information

signalword Warning
pcodes P271;P261;P280
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart