APO414281910 Apollo Scientific Ltd

1-Acetylpiperidine-3-carboxylic acid

  • MDL number MFCD07348751
  • Cas Number : 2637-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-132
Synonym 1-Ethanoylpiperidine-3-carboxylic acid, 1-Acetyl-3-carboxypiperidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-acetylpiperidine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-acetylpiperidine-3-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart