APO414281959 Apollo Scientific Ltd

6-Bromo-1,3-dihydro-2H-imidazo[4,5-b]pyridin-2-one

  • MDL number MFCD00763285
  • Cas Number : 148038-83-9
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp >350
Iupac Name 6-bromo-1H-imidazo[4,5-b]pyridin-2-ol
Inchi Code 1S/C6H4BrN3O/c7-3-1-4-5(8-2-3)10-6(11)9-4/h1-2H,(H2,8,9,10,11)
Inchi Key VHCRLTJPUNUZRN-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point >350
Product Link https://store.apolloscientific.co.uk/product/6-bromo-13-dihydro-2h-imidazo45-bpyridin-2-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-13-dihydro-2h-imidazo45-bpyridin-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart