APO414282001 Apollo Scientific Ltd

6-Chloroisatoic anhydride

  • MDL number MFCD09032504
  • Cas Number : 20829-96-3
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloroisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloroisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart