APO414282012 Apollo Scientific Ltd

4-(Trifluoromethyl)isatoic anhydride

  • MDL number MFCD21946140
  • Cas Number : 97928-01-3
  • Molecular Weight : 231.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-242
Synonym 7-(Trifluoromethyl)-2H-3,1-benzoxazine-2,4(1H)-dione, 1,4-Dihydro-2,4-dioxo-7-(trifluoromethyl)-2H-3,1-benzoxazine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylisatoic-anhydride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylisatoic-anhydride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart