APO414282016 Apollo Scientific Ltd

6-Bromoisatoic anhydride

  • MDL number MFCD15526667
  • Cas Number : 77603-45-3
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1H-benzo[d][1,3]oxazine-2,4-dione
Inchi Code 1S/C8H4BrNO3/c9-4-2-1-3-5-6(4)7(11)13-8(12)10-5/h1-3H,(H,10,12)
Inchi Key YELXVMUMHRDCKE-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromoisatoic-anhydride

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/6-bromoisatoic-anhydride
pictograms GHS05;GHS08;GHS07
hstatements H302+H312+H332;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart