APO414282165 Apollo Scientific Ltd

2-Amino-4-bromo-3-fluorobenzoic acid

  • MDL number MFCD27664889
  • Cas Number : 1416013-62-1
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-bromo-3-fluorobenzoic acid
Inchi Code 1S/C7H5BrFNO2/c8-4-2-1-3(7(11)12)6(10)5(4)9/h1-2H,10H2,(H,11,12)
Inchi Key SFRJQFICUQFDFU-UHFFFAOYSA-N

Description

Synonym 3-Bromo-6-carboxy-2-fluoroaniline, 4-Bromo-3-fluoroanthranilic acid
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-bromo-3-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-bromo-3-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart