APO414282168 Apollo Scientific Ltd

2,3-Difluoro-4-nitroaniline

  • MDL number MFCD26960577
  • Cas Number : 1781140-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60294

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=60294
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart