APO414282192 Apollo Scientific Ltd

4-Chloro-3-methoxyphenol

  • MDL number MFCD16998742
  • Cas Number : 18113-07-0
  • Molecular Weight : 158.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-methoxyphenol
Inchi Code 1S/C7H7ClO2/c1-10-7-4-5(9)2-3-6(7)8/h2-4,9H,1H3
Inchi Key NKFRBXPBRPYULV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-methoxyphenol

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-methoxyphenol
pictograms GHS05;GHS07
hstatements H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart