APO414282208 Apollo Scientific Ltd

2-Bromo-5-iodophenylacetonitrile

  • MDL number MFCD18391771
  • Cas Number : 1261517-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-iodobenzyl cyanide, (2-Bromo-5-iodophenyl)acetonitrile
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59486

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59486
pictograms GHS06
hstatements H301;H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart