APO414282216 Apollo Scientific Ltd

5-Bromo-4-fluoro-2-nitroanisole

  • MDL number MFCD16658620
  • Cas Number : 330794-02-0
  • Molecular Weight : 250.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-5-fluoro-2-methoxynitrobenzene, 1-Bromo-2-fluoro-5-methoxy-4-nitrobenzene, 5-Bromo-4-fluoro-2-nitrophenyl methyl ether
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-4-fluoro-2-nitroanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-4-fluoro-2-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart