APO414282218 Apollo Scientific Ltd

2-Methyl-3-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD09832279
  • Cas Number : 1000546-18-8
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 105-108
Iupac Name [2-methyl-3-(trifluoromethyl)phenyl]acetic acid
Inchi Code 1S/C10H9F3O2/c1-6-7(5-9(14)15)3-2-4-8(6)10(11,12)13/h2-4H,5H2,1H3,(H,14,15)
Inchi Key FQVYJURFBWJJPW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 105-108
Product Link https://store.apolloscientific.co.uk/product/2-methyl-3-trifluoromethylphenylacetic-acid

Safety Information

signalword Warning
pcodes P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-methyl-3-trifluoromethylphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart