APO414282298 Apollo Scientific Ltd

1-Benzyloxycarbonyl-3-azetidinecarboxylic acid methyl ester

  • MDL number MFCD11110708
  • Cas Number : 757239-60-4
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl 3-methyl 1,3-azetidinedicarboxylate
Inchi Code 1S/C13H15NO4/c1-17-12(15)11-7-14(8-11)13(16)18-9-10-5-3-2-4-6-10/h2-6,11H,7-9H2,1H3
Inchi Key VMBHUIMZBSWWCN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-benzyloxycarbonyl-3-azetidinecarboxylic-acid-methyl-ester

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/1-benzyloxycarbonyl-3-azetidinecarboxylic-acid-methyl-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart