APO414282301 Apollo Scientific Ltd

Methyl 5-bromopyrazine-2-carboxylate

  • MDL number MFCD09991715
  • Cas Number : 210037-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-(methoxycarbonyl)pyrazine, 2-Bromo-5-(methoxycarbonyl)-1,4-diazine
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52203

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52203
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart