APO414282345 Apollo Scientific Ltd

2-(Trifluoromethyl)pyrimidin-5-ol

  • MDL number MFCD10696944
  • Cas Number : 100991-09-1
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(trifluoromethyl)-5-pyrimidinol
Inchi Code 1S/C5H3F3N2O/c6-5(7,8)4-9-1-3(11)2-10-4/h1-2,11H
Inchi Key QPAXXIOBBIIATH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylpyrimidin-5-ol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylpyrimidin-5-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart