APO414282349 Apollo Scientific Ltd

5-Fluoropyrimidin-4-amine

  • MDL number MFCD00223685
  • Cas Number : 811450-26-7
  • Molecular Weight : 113.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-4-pyrimidinamine
Inchi Code 1S/C4H4FN3/c5-3-1-7-2-8-4(3)6/h1-2H,(H2,6,7,8)
Inchi Key PKZMNMQCFNJXEH-UHFFFAOYSA-N

Description

Flash Point 222.7/760mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoropyrimidin-4-amine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoropyrimidin-4-amine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart