APO414282370 Apollo Scientific Ltd

1-Methylpiperazin-2-one

  • MDL number MFCD00102572
  • Cas Number : 59702-07-7
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 134-137/0.5mm
Product Link https://store.apolloscientific.co.uk/product/1-methylpiperazin-2-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylpiperazin-2-one
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart