APO414282376 Apollo Scientific Ltd

2-(Chloromethyl)-1,3-oxazole

  • MDL number MFCD04114926
  • Cas Number : 185246-17-7
  • Molecular Weight : 117.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(chloromethyl)-1,3-oxazole
Inchi Code 1S/C4H4ClNO/c5-3-4-6-1-2-7-4/h1-2H,3H2
Inchi Key NAOPGVBLRHCPHI-UHFFFAOYSA-N

Description

Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloromethyl-13-oxazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloromethyl-13-oxazole
pictograms GHS07
hstatements H302;H315;H319;H412
Availability details for
Quantity
  • Availability
  • Add to Cart