APO414282586 Apollo Scientific Ltd

3-Bromo-2-chloro-6-fluorobenzaldehyde

  • MDL number MFCD11110260
  • Cas Number : 1114809-11-8
  • Molecular Weight : 237.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-chloro-6-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-chloro-6-fluorobenzaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart