APO414282781 Apollo Scientific Ltd

1-tert-Butyl 2-methyl 3-oxopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD24369719
  • Cas Number : 194924-96-4
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butyl) 2-methyl 3-oxopyrrolidine-1,2-dicarboxylate
Inchi Code 1S/C11H17NO5/c1-11(2,3)17-10(15)12-6-5-7(13)8(12)9(14)16-4/h8H,5-6H2,1-4H3
Inchi Key SZAMWQSLASHJAF-UHFFFAOYSA-N

Description

Synonym 1-(tert-Butoxycarbonyl)-2-(methoxycarbonyl)-3-oxopyrrolidine, 3-Oxo-DL-proline methyl ester, N-BOC protected
Flash Point 140/0.375mm
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-tert-butyl-2-methyl-3-oxopyrrolidine-12-dicarboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-tert-butyl-2-methyl-3-oxopyrrolidine-12-dicarboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart