APO414282817 Apollo Scientific Ltd

5-Hydroxy-1H-pyrazole

  • MDL number MFCD11227225
  • Cas Number : 137-45-1
  • Molecular Weight : 84.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-179
Synonym 2,3-Dihydro-1H-pyrazol-3-one, 2H-Pyrazol-3-ol, 1H-Pyrazol-3-ol, 1H-Pyrazol-5-ol, 1,2-Dihydro-3H-pyrazol-3-one
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-hydroxy-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxy-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart