APO414282948 Apollo Scientific Ltd

5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride

  • Cas Number : 175277-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Trifluoromethy)pyridine-2-amidine hydrochloride
Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52660

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=52660
Availability details for
Quantity
  • Availability
  • Add to Cart