APO414283002 Apollo Scientific Ltd

2,3,5-Trichlorobenzaldehyde

  • MDL number MFCD00010860
  • Cas Number : 56961-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Synonym 2,3,5-Trichlorobenzene carboxaldehyde
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52706

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52706
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart