APO414283043 Apollo Scientific Ltd

2-Chloro-4-fluoro-5-iodoanisole

  • MDL number MFCD22689435
  • Cas Number : 153122-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Synonym 1-Chloro-5-fluoro-4-iodo-2-methoxybenzene, 2-Chloro-4-fluoro-5-iodophenyl methyl ether
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53145

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53145
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart