APO414283068 Apollo Scientific Ltd

Methyl 2-bromo-3-methylbenzoate

  • MDL number MFCD00144762
  • Cas Number : 131001-86-0
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-3-methylbenzoate
Inchi Code 1S/C9H9BrO2/c1-6-4-3-5-7(8(6)10)9(11)12-2/h3-5H,1-2H3
Inchi Key QAOFGUXVDAZKBW-UHFFFAOYSA-N

Description

Synonym 2-Bromo-3-methylbenzoic acid methyl ester
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-3-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-3-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart