APO414283167 Apollo Scientific Ltd

6-Chloro-3-hydroxy-4-methylpyridazine

  • MDL number MFCD24539279
  • Cas Number : 1834-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Chloro-4-methylpyridazin-3-ol, 6-Chloro-4-methylpyridazin-3(2H)-one
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-chloro-3-hydroxy-4-methylpyridazine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-3-hydroxy-4-methylpyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart