APO414283169 Apollo Scientific Ltd

4-Amino-3,6-dichloropyridazine

  • MDL number MFCD01647253
  • Cas Number : 823-58-5
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,6-dichloro-4-pyridazinylamine
Inchi Code 1S/C4H3Cl2N3/c5-3-1-2(7)4(6)9-8-3/h1H,(H2,7,8)
Inchi Key HODYDVHWWMTUEL-UHFFFAOYSA-N

Description

Synonym 3,6-Dichloropyridazin-4-amine, 4-Amino-3,6-dichloro-1,2-diazine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-36-dichloropyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-36-dichloropyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart