APO414283198 Apollo Scientific Ltd

3-Chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD17168283
  • Cas Number : 1237838-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53730

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=53730
Availability details for
Quantity
  • Availability
  • Add to Cart