APO414284118 Apollo Scientific Ltd

(1R,2S)-2-(Methoxycarbonyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD18374521
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl (1R,2R)-2-carboxycyclohexane-1-carboxylate, (1R,2S)-1-Carboxy-2-(methoxycarbonyl)cyclohexane
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1r2s-2-methoxycarbonylcyclohexane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1r2s-2-methoxycarbonylcyclohexane-1-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart