APO414284221 Apollo Scientific Ltd

3-Amino-N-(tert-butoxycarbonyl)-D-alanine

  • MDL number MFCD01632072
  • Cas Number : 76387-70-7
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-3-amino-2-[(tert-butoxycarbonyl)amino]propanoic acid
Inchi Code 1S/C8H16N2O4/c1-8(2,3)14-7(13)10-5(4-9)6(11)12/h5H,4,9H2,1-3H3,(H,10,13)(H,11,12)/t5-/m1/s1
Inchi Key KRJLRVZLNABMAT-RXMQYKEDSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-n-tert-butoxycarbonyl-d-alanine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-n-tert-butoxycarbonyl-d-alanine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart