APO414284285 Apollo Scientific Ltd

1-Benzofuran-7-ylmethanol

  • MDL number MFCD19441765
  • Cas Number : 209256-55-3
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-benzofuran-7-ylmethanol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-benzofuran-7-ylmethanol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart