APO414284289 Apollo Scientific Ltd

3-(Chloromethyl)-2-hydroxy-5-methylbenzaldehyde

  • MDL number MFCD03055723
  • Cas Number : 192819-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-chloromethyl-2-hydroxy-5-methylbenzaldehyde

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloromethyl-2-hydroxy-5-methylbenzaldehyde
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart