APO414284331 Apollo Scientific Ltd

N,N-Dimethyloctanamide

  • MDL number MFCD00043763
  • Cas Number : 1118-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -21
Density 0.881
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 187/100mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=56800

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=56800
pictograms GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart