APO414284347 Apollo Scientific Ltd

1-Bromo-3,5-dinitrobenzene

  • MDL number MFCD00156596
  • Cas Number : 18242-39-2
  • Molecular Weight : 247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-35-dinitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-35-dinitrobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart