APO414284496 Apollo Scientific Ltd

2-(Hydroxymethyl)piperazine, N1-BOC protected

  • MDL number MFCD07772091
  • Cas Number : 205434-75-9
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 2-(hydroxymethyl)piperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-2-(hydroxymethyl)piperazine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxymethylpiperazine-n1-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxymethylpiperazine-n1-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart