APO414284514 Apollo Scientific Ltd

Ethyl 3-hydroxycyclobutane-1-carboxylate

  • MDL number MFCD09758972
  • Cas Number : 17205-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Ethoxycarbonyl)cyclobutan-1-ol
Purity 95
Density 1.0727
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 120/10mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=57072

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=57072
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart