APO414284524 Apollo Scientific Ltd

tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate

  • MDL number MFCD09839640
  • Cas Number : 896464-16-7
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate
Inchi Code 1S/C12H22N2O2/c1-11(2,3)16-10(15)14-6-4-12(5-7-14)8-13-9-12/h13H,4-9H2,1-3H3
Inchi Key NRADOPGBTAJXKB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-27-diazaspiro35nonane-7-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-27-diazaspiro35nonane-7-carboxylate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart