APO414285496 Apollo Scientific Ltd

2-Bromo-6-chlorophenol

  • MDL number MFCD09907875
  • Cas Number : 2040-88-2
  • Molecular Weight : 207.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 55-56
Iupac Name 2-bromo-6-chlorophenol
Inchi Code 1S/C6H4BrClO/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
Inchi Key LUECOFFMVWPWDR-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 55-56
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-chlorophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-chlorophenol
pictograms GHS07
hstatements H302;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart