APO414285524 Apollo Scientific Ltd

3-Bromo-2-fluoro-6-nitrotoluene

  • MDL number MFCD26407497
  • Cas Number : 1807209-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-fluoro-3-methyl-4-nitrobenzene, 4-Bromo-3-fluoro-2-methylnitrobenzene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=58695

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=58695
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart