APO414285966 Apollo Scientific Ltd

3-([Pentafluorothio)benzeneboronic acid, pinacol ester

  • MDL number MFCD20489193
  • Cas Number : 1286278-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,4,5,5-Tetramethyl-2-(m-pentafluorosulfanylbenzene)-1,3,2-dioxaborolane
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59796

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59796
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart