APO414285967 Apollo Scientific Ltd

4-(Aminomethyl)phenylsulphur pentafluoride

  • MDL number MFCD06213675
  • Cas Number : 771573-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Pentafluorothio)benzylamine, [4-(Pentafluorosulphanyl)phenyl]methylamine
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59765

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59765
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart