APO414286003 Apollo Scientific Ltd

Bicyclo[1.1.1]pentane-1-carboxylic acid

  • MDL number MFCD19228106
  • Cas Number : 22287-28-1
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 125-127/45mm
Product Link https://store.apolloscientific.co.uk/product/bicyclo111pentane-1-carboxylic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/bicyclo111pentane-1-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart